Learn More
Thermo Scientific™ Hypersil™ PREP HS Si HPLC Columns
Separate nonpolar and moderately polar compounds with high loading capacity Hypersil PREP HS Si, fully scalable from analytical to prep for USP-L3 applications.
$180.30 - $20190.00
Specifications
Particle Size | 5 μm |
---|---|
pH | 2 to 7 |
Pore Size | 100 Å |
Surface Area | 350 m²/g |
Temperature | 60°C |
Catalog Number | Mfr. No. | Column Format | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Column Format | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
03-253-082
|
Thermo Scientific™
41405018823A |
Guard Column | 10 mm | 8 mm |
Each for $180.30
|
|
|||
03-253-083
|
Thermo Scientific™
41405019023A |
Guard Column | 10 mm | 20 mm |
Each for $313.50
|
|
|||
03-253-085
|
Thermo Scientific™
41405019323A |
Guard 1/16 in. | 10 mm | 30 mm |
Each for $379.00
|
|
|||
03-253-709
|
Thermo Scientific™
41405019323B |
Guard 1/8 in. | 10 mm | 30 mm |
Each for $379.00
|
|
|||
03-253-700
|
Thermo Scientific™
41405029323A |
Guard 1/16 in. | 20 mm | 30 mm |
Each for $625.00
|
|
|||
03-253-710
|
Thermo Scientific™
41405029323B |
Guard 1/8 in. | 20 mm | 30 mm |
Each for $625.00
|
|
|||
03-253-062
|
Thermo Scientific™
41405052130 |
Preparative Column, Standard | 2 mm | 50 mm |
Each for $512.50
|
|
|||
03-253-066
|
Thermo Scientific™
41405054630 |
Preparative Column, Standard | 4.6 mm | 50 mm |
Each for $592.00
|
|
|||
03-253-070
|
Thermo Scientific™
41405059070A |
Preparative Column, Standard | 10 mm | 50 mm |
Each for $1,390.00
|
|
|||
03-253-071
|
Thermo Scientific™
41405059270A |
Preparative Column, Standard | 20 mm | 50 mm |
Each for $1,475.00
|
|
|||
03-253-701
|
Thermo Scientific™
41405059270XL |
Preparative Column, XtendedLife | 25 mm | 50 mm |
Each for $5,520.00
|
|
|||
03-253-063
|
Thermo Scientific™
41405102130 |
Preparative Column, Standard | 2 mm | 100 mm |
Each for $659.50
|
|
|||
03-253-067
|
Thermo Scientific™
41405104630 |
Preparative Column, Standard | 4.6 mm | 100 mm |
Each for $728.00
|
|
|||
03-253-072
|
Thermo Scientific™
41405109070A |
Preparative Column, Standard | 10 mm | 100 mm |
Each for $1,475.00
|
|
|||
03-253-073
|
Thermo Scientific™
41405109270A |
Preparative Column, Standard | 20 mm | 100 mm |
Each for $2,800.00
|
|
|||
03-253-702
|
Thermo Scientific™
41405109270XL |
Preparative Column, XtendedLife | 25 mm | 100 mm |
Each for $7,350.00
|
|
|||
03-253-064
|
Thermo Scientific™
41405152130 |
Preparative Column, Standard | 2 mm | 150 mm |
Each for $714.50
|
|
|||
03-253-068
|
Thermo Scientific™
41405154630 |
Preparative Column, Standard | 4.6 mm | 150 mm |
Each for $812.00
|
|
|||
03-253-074
|
Thermo Scientific™
41405159070A |
Preparative Column, Standard | 10 mm | 150 mm |
Each for $1,687.00
|
|
|||
03-253-075
|
Thermo Scientific™
41405159270A |
Preparative Column, Standard | 20 mm | 150 mm |
Each for $3,340.00
|
|
|||
03-253-703
|
Thermo Scientific™
41405159270XL |
Preparative Column, XtendedLife | 25 mm | 150 mm |
Each for $8,020.00
|
|
|||
03-253-076
|
Thermo Scientific™
41405159370A |
Preparative Column, Standard | 30 mm | 150 mm |
Each for $5,460.00
|
|
|||
03-253-704
|
Thermo Scientific™
41405159370XL |
Preparative Column, XtendedLife | 30 mm | 150 mm |
Each for $9,930.00
|
|
|||
03-253-077
|
Thermo Scientific™
41405159570A |
Preparative Column, Standard | 50 mm | 150 mm |
Each for $10,850.00
|
|
|||
03-253-705
|
Thermo Scientific™
41405159570XL |
Preparative Column, XtendedLife | 50 mm | 150 mm |
Each for $17,100.00
|
|
|||
03-253-065
|
Thermo Scientific™
41405252130 |
Preparative Column, Standard | 2 mm | 250 mm |
Each for $836.50
|
|
|||
03-253-069
|
Thermo Scientific™
41405254630 |
Preparative Column, Standard | 4.6 mm | 250 mm |
Each for $965.00
|
|
|||
03-253-078
|
Thermo Scientific™
41405259070A |
Preparative Column, Standard | 10 mm | 250 mm |
Each for $1,920.00
|
|
|||
03-253-079
|
Thermo Scientific™
41405259270A |
Preparative Column, Standard | 20 mm | 250 mm |
Each for $4,020.00
|
|
|||
03-253-706
|
Thermo Scientific™
41405259270XL |
Preparative Column, XtendedLife | 25 mm | 250 mm |
Each for $7,860.00
|
|
|||
03-253-080
|
Thermo Scientific™
41405259370A |
Preparative Column, Standard | 30 mm | 250 mm |
Each for $6,470.00
|
|
|||
03-253-707
|
Thermo Scientific™
41405259370XL |
Preparative Column, XtendedLife | 30 mm | 250 mm |
Each for $10,490.00
|
|
|||
03-253-081
|
Thermo Scientific™
41405259570A |
Preparative Column, Standard | 50 mm | 250 mm |
Each for $12,210.00
|
|
|||
03-253-708
|
Thermo Scientific™
41405259570XL |
Preparative Column, XtendedLife | 50 mm | 250 mm |
Each for $20,190.00
|
|
|||
Description
Easily scale up your analytical method and achieve maximum retention and loading using Thermo Scientific™ Hypersil™ PREP HS Si preparative HPLC columns. Packed with a bare silica, the high surface area particles with a 100Å pore size are made for normal phase chromatography of nonpolar and moderately polar organic compounds. Our preparative HPLC columns come in a variety of dimensions including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed.
Normal phase chromatography
- Silica to separate nonpolar and moderately polar organic compounds.
- USP L3 - high surface area particle with 100Å pore size.
- pH stable from 2 to .7
- Maximum temperature of 60°C.
Preparative HPLC Column Hardware Options - based on column inner diameter (i.d.) and column performance needs.
Standard Threaded and Standard Flange Hardware
- Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use.
- Standard threaded hardware for column i.d. < 40mm.
- Standard flange hardware for column i.d. ≥ 40mm.
- Standard hardware can be modified on request to be compatible with SFC.
XtendedLife Hardware
- Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly.
- Compatible with SFC and is available in dimensions up to 50mm i.d.
Preparative Guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.
Customer Specific Orders
- If there is a Thermo Scientific stationary phase you are currently using and would like to obtain in a preparative scale not shown or would like to discuss a specific column dimension, we would be happy to discuss your requirements.
- Please contact your local sales representative for Customer Specific Orders.
- Custom built columns are not eligible for returns.
Specifications
5 μm | |
100 Å | |
60°C | |
Silica | |
0% | |
No |
2 to 7 | |
350 m²/g | |
L3 | |
Spherical, Fully Porous, Ultrapure Silica | |
Mixed Mode | |
Normal Phase and HILIC |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.