Learn More
Perkin Elmer US LLC HyperDSC Aluminum Sample Pans - Qty. 100
Supplier: Perkin Elmer US LLC N5203115
These thin ultra-light foil pans can be used to run HyperDSC methods on solid samples. The thin material provides optimal thermal contact between the sample and the sensor. The sample wrap reduces thermal gradients while scanning at fast rates.