Learn More
EUROTROL HYPERBARIC QC 4 AMPULES/BOX
Supplier: EUROTROL 164.002.004
Extremely high pO2 to extend reportable range. Room temperature storage and stability. Shelf life of 3 years. Use with CueSee Online, free data analysis/peer comparison service.