Learn More
Hydroxyethyl Cellulose, 5,000 cP, NF, 50 kg, Spectrum Chemical
Click to view available options
Quantity: 50 kg
Description
Hydroxyethyl Cellulose, 2,000 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent for aqueous pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Specifications
Quantity | 50 kg |