Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STERIS™ Hydrogen Peroxide WFI Sterile Solution
A low residue cleaner with minimal organic carbon
Supplier: STERIS™ 1S0596WR
This item is not returnable.
View return policy
Description
- Made with Hydrogen Peroxide and USP-quality Water For Injection (WFI)
- Formulated to be a low residue cleaner with minimal organic carbon
- Filtered, and filled in an ISO Class 5 aseptic environment to minimize the potential of particles being introduced during use
- Each package is double bagged for easy entry into controlled environments
- Includes an extensive documentation package to meet validation objectives
- Supported by the STERIS Technical Services team for individualized application and validation assistance
- Each lot is documented by a Certificate of Analysis that includes sterility results
Specifications
Disinfectant | |
Hydrogen Peroxide and USP-quality Water For Injection (WFI) | |
Trigger Spray Bottle | |
Removing Organic and Inorganic Residue From Surfaces | |
32 oz. |
Sterile | |
0.06% | |
Filtered | |
12/Cs. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction