Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYDRANAL™ - NEXTGEN Solvent E-FI, Honeywell Fluka™
Medium for volumetric, two-component Karl Fischer titrations
$739.00
Specifications
Density | 0.895 g/cm3 |
---|---|
Chemical Name or Material | HYDRANAL™ - NEXTGEN Solvent E-FI |
Color | Colorless |
CAS | 64-17-5 |
pH | 5 to 6 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
60-049-488
|
Honeywell Chemicals
344316X1L |
6 x 1 L |
Case of 6 for $739.00
|
|
|||||
Description
- Ethanol-based
- Free of Imidazole
- Free of CMR Substances
Specifications
0.895 g/cm3 | |
Colorless | |
5 to 6 | |
≤80% | |
-115°C | |
UN1170 |
HYDRANAL™ - NEXTGEN Solvent E-FI | |
64-17-5 | |
≥55% | |
Glass Bottle | |
Liquid | |
Completely Miscible in Water |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title