Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Hyaluronan Quantikine ELISA Kit
Hyaluronan Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
$736.60 - $4199.20
Specifications
Conjugate | HRP |
---|---|
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates |
Specificity | 35 kDa Hyaluronan in Human, Mouse, Rat, Canine, and Porcine Samples |
For Use With (Application) | ELISA |
Description
The Hyaluronan Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies multi-species Hyaluronan in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).Specifications
HRP | |
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates | |
35 kDa Hyaluronan in Human, Mouse, Rat, Canine, and Porcine Samples | |
ELISA |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title