Learn More
Techcyte Human Trichrome O&P Fecal Analysis
Human Trichrome Ova and Parasite Fecal Analysis: with a slide-level sensitivity of 98.9% and specificity of 98.1%, our fecal ova and parasite solution is five times more sensitive than manual examination.
Supplier: Techcyte HFTPAYG
Description
- Involves scanning a human fecal trichrome-stained microscope slide and analyzing it using the Techcyte HFT algorithm
- The algorithm presents predicted images for human fecal parasites classes to the user for diagnosis
Specifications
Detection Software | |
Analyzing Data |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Techcyte is CE-IVDD marked in Europe, and for investigational use only in the US.
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'