Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human PDGF-AB Quantikine ELISA Kit

Human PDGF-AB Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

$736.60 - $4199.20

Specifications

Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 3.83pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type ELISA Kit
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Quantity Sufficient For Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity Sufficient For Price Quantity & Availability  
DHD00C
View Documents
R&D Systems™
DHD00C
ELISA Kit 1 Kit 1 x 96-well plate
Each for $736.60
Add to Cart
 
SHD00C
View Documents
R&D Systems™
SHD00C
ELISA Kit 1 Kit 6 x 96-well plates
Each for $4,199.20
Add to Cart
 
Description

Description

The Human PDGF-AB Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human PDGF-AB in cell culture supernates (100 ul), serum (10 ul), platelet-poor edta plasma (100 ul), platelet-poor heparin plasma (100 ul).
Specifications

Specifications

15.60 - 1000pg/mL
HRP
ELISA Kit
Natural and Recombinant Human PDGF-AB
ELISA
Cell Culture Supernates (100uL),Serum (10uL),Platelet-poor EDTA Plasma (100uL),Platelet-poor Heparin Plasma (100uL)
3.83pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Cell Culture Supernatant, Platelet-poor Plasma (EDTA, Heparin), Serum
Human
1 Kit
Store the unopened product at 2° to 8°C. Do not use past expiration date.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit