Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human IL-2 DuoSet ELISA
Human IL-2 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
$523.50 - $950.50
Specifications
Product Type | ELISA Development Kit |
---|---|
Quantity | 1 Kit |
Species | Human |
Conjugate | Biotin |
Assay Range | 15.60 to 1000 pg/mL |
Description
The Human IL-2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-2.Specifications
ELISA Development Kit | |
Human | |
15.60 to 1000 pg/mL | |
ELISA | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
1 Kit | |
Biotin | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
3558 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title