Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human IL-10 Quantikine ELISA Kit
Human IL-10 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
$736.60 - $4199.20
Specifications
Assay Range | 7.80 to 500 pg/mL |
---|---|
Assay Sensitivity | 3.9 pg/mL |
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
Product Type | Solid Phase Sandwich ELISA |
Description
The Human IL-10 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-10 in cell culture supernates (200 ul), serum (200 ul), edta plasma (200 ul), heparin plasma (200 ul), citrate plasma (200 ul).Specifications
7.80 to 500 pg/mL | |
HRP | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Human IL-10 | |
ELISA | |
IL10 | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10 mgC126451, interleukin 10, interleukin-10, TGIF |
3.9 pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
Cell Culture Supernatant, Citrate, Plasma (EDTA, Citrate, Heparin), Serum | |
Human | |
3586 | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
IL-10 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title