Learn More
MilliporeSigma™ Human IgG2, Myeloma, Isotype Control,
Human Isotype Control Antibody
Supplier: MilliporeSigma™ 4001222MG
Description
Specifically detects IgG2, Myeloma Clone: in samples, and it is validated for Isotype Control
Specifications
IgG2, Myeloma | |
In 150mM NaCl, 20mM phosphate buffer, pH 7.4. | |
2 mg | |
Isotype Control (Primary) | |
−20°C |
Unconjugated | |
Human | |
RUO | |
Native IgG2 from human myeloma plasma. | |
Liquid |