Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human IgG Fc ELISA Kit (Colorimetric)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
CLU Colorimetric ELISA Kit is useful in the study of Apolipoprotein
Supplier: Novus Biologicals™ NBP2600681KIT
Description
The Human IgG Fc ELISA Kit (Colorimetric) measures human IgG Fc in serum, plasma.Specifications
0.027 to 20μg/mL | |
ELISA Kit (Colorimetric) | |
Detection of human Fcy | |
ELISA | |
1 Kit | |
Immunology | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. |
0.024μg/mL | |
Plasma, Serum | |
Human | |
Assay plate (12 x 8 coated Microwells), Standard (Freeze dried), Biotin-antibody (100X concentrate), RP-avidin (100X concentrate), Biotin-antibody Diluents, HRP-avidin Diluents, Sample Diluents, Wash Buffer (25X concentrate), TMB Substrate, Stop Solution, Adhesive Strip (For 96 wells), Instruction manual | |
RUO | |
50 to 100 μL | |
IgG Fc |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction