Learn More
MilliporeSigma™ Human IgA2, Myeloma, Isotype Control,
Human Isotype Control Antibody
Supplier: MilliporeSigma™ 400110500UG
Description
Specifically detects IgA2, Myeloma Clone: in samples, and it is validated for Isotype Control
Specifications
IgA2, Myeloma | |
In 150mM NaCl, 10mM phosphate buffer, pH 7.4. | |
500 μg | |
Isotype Control (Primary) | |
−20°C |
Unconjugated | |
Human | |
RUO | |
Native IgA2 from human myeloma plasma. | |
Liquid |