Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human IFN-gamma Quantikine ELISA Kit
solid phase ELISA designed to measure human IFN-gamma levels in cell culture supernates, serum, and plasma.
$736.60 - $4199.20
Specifications
Assay Range | 15.60 to 1000 pg/mL |
---|---|
Assay Sensitivity | 5.69 pg/mL |
Sample Type | Cell Culture Supernatant, Plasma (EDTA), Heparin Plasma, Serum |
Specificity | Natural and Recombinant Human IFN-gamma |
Target Species | Human |
Specifications
15.60 to 1000 pg/mL | |
Cell Culture Supernatant, Plasma (EDTA), Heparin Plasma, Serum | |
Human | |
3458 | |
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma |
5.69 pg/mL | |
Natural and Recombinant Human IFN-gamma | |
ELISA | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
IFN-gamma |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title