Learn More
Human IFN-γ ELISPOT Pair, BD
Convenient set of unlabelled anti-IFN-γ capture antibody and a biotinylated anti-IFN-γ detection antibody
Supplier:  BD Biosciences 551873
Catalog No. BDB551873
Add to Cart
Includes: Human IFN-γ ELISPOT Capture Antibody (no azide/low endotoxin), Human IFN-γ ELISPOT Detection Antibody (biotinylated) and a Certificate of Analysis that provides lot-specific optimal reagent concentrations