Learn More
PBL Assay Science VeriKine Human Inteferon Beta (IFN-Beta, IFNb), Tissue Culture Media (TCM) (TCM) [5x96 well plate]
Supplier: PBL Assay Science 414102
VeriKine Human Interferon Beta ELISA Kit is manufactured by PBL Assay Science (Catalog #41410-2) for research use only (RUO) and packaged as a 5x96-well plate. This IFNb ELISA is Tissue Culture Media (TCM) compatible, quantifies Hu IFN-Beta, and has an assay range of 50-4000 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 8%.