Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human GLP-1R APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2814A
This item is not returnable.
View return policy
Description
GLP-1R Monoclonal specifically detects GLP-1R in Human samples. It is validated for Flow Cytometry.Specifications
GLP-1R | |
Monoclonal | |
APC | |
NP_002053 | |
GLP1R | |
NS0 mouse myeloma cell line transfected with human GLP-1R Arg24-Ser463 Accession # NP_002053 | |
100 Tests | |
2740 | |
Human | |
Purified |
Flow Cytometry | |
197920 | |
Flow Cytometry 10 uL/10^6 cells | |
GLP-1 receptor, GLP1R, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Stains human GLP-1R transfectants but not irrelevant transfectants in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction