Learn More
Cytiva Human Fab Capture Kit, type 2
Supplier: Cytiva 29234601
Reagents for capture of human Fab fragments as ligands in biomolecular interaction analysis.Designed for use with Biacore 8K (larger kit component volumes than in Human Fab Capture Kit) but can be used with all other Biacore systems.