Learn More
Quantimetrix Dropper Urine Chemistry Control, Level 2
Supplier: Quantimetrix 143231
•Easy-to-use dropper bottles•Liquid, made with human urine•2-year shelf life & open vial stability from date of manufacture when stored at 2ºC–8ºCUse Dropper Urine Chemistry Control to monitor the performance of routine and stat urine chemistry assay methods. Drop the control directly into the instrument sample cup. The easy-to-use dropper bottle provides simple dispensing and maximum use. Dropper Urine Chemistry Control is designed for use with most major chemistry analyzers including: Siemens Advia, Beckman Coulter AU Series, Siemens Dimension, Abbott Architect and Roche Cobas, as well as for the Pyrogallol Red microprotein method and osmolality (freezing point method).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.