Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Contactin-2/TAG1 (NP_005067) VersaClone cDNA
CNTN2 contactin 2 Protein
Supplier: R&D Systems™ RDC1922
This item is not returnable.
View return policy
Description
The Human Contactin-2/TAG1 (NP_005067) VersaClone cDNA clone contains the complete ORF for Contactin-2/TAG1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Contactin-2/TAG1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Specifications
cDNA | |
NP_005067 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
6900 (Human) | |
Human |
hCNTN2 | |
10 μg at 0.2 μg/μL | |
3136bp | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction