Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human ARID3A Full-length ORF (AAH60828.1, 1 a.a. - 593 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00001820P01
Description
This gene encodes a member of the ARID (AT-rich interaction domain) family of DNA binding proteins. It was found by homology to the Drosophila dead ringer gene, which is important for normal embryogenesis. Other ARID family members have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation, and possibly in chromatin structure modification. [provided by RefSeq]
Sequence: MKLQAVMETLLQRQQRARQELEARQQLPPDPPAAPHGRARAAPDEDREPESARMQRAQMAALAAMRAAAAGLGHPASPGGSEDGPPGSEEEDAAREGTPGSPGRGREGPGEEHFEDMASDEDMKPKWEEEEMEEDLGEDEEEEEEDYEDEEEEEDEEGLGPPGPASLGTTALFPRKAQPPQAFRGDGVPRVLGGQERPGPGPAHPGGAAHVAPQLQPPDHGDWTYEEQFKQLYELDGDPKRKEFLDDLFSFMQKRGTPVNRIPIMAKQVLDLFMLYVLVTEKGGLVEVINKKLWREITKGLNLPTSITSAAFTLRTQYMEYLYPYECEKRGLSNPNELQAAIDSNRREGRRQSFGGSLFAYSPGGAHGMLSSPKLPVSSLGLAASTNGSSITPAPKIKKEEDSAIPITVPGRLPVSLAGHPVVAAQAAAVQAAAAQAAVAAQAAALEQLREKLESAEPPEKKMALVADEQQRLMQRALQQNFLAMAAQLPMSIRINSQASESRQDSAVNLTGTNGSNSISMSVEINGIMYTGVLFAQPPAPTPTSAPNKGGGGGGSSSSNAGGRGGNTGTSGGQAGPAGLSTPSTSTSNNSLPSpecifications
AAH60828.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
89.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
RUO | |
ARID3A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
1820 | |
ARID3A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BRIGHT/DRIL1/DRIL3/E2FBP1 | |
ARID3A | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction