Learn More
Abnova™ Human ARHGEF6 Full-length ORF (NP_004831.1, 1 a.a. - 776 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_004831.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9459 |
Molecular Weight (g/mol) | 113.9kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-935-556
|
Abnova™
H00009459P01S |
10 ug |
Each for $499.00
|
|
|||||
89-935-557
|
Abnova™
H00009459P01L |
25 ug |
Each for $769.00
|
|
|||||
Description
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein belongs to a family of cytoplasmic proteins that activate the Ras-like family of Rho proteins by exchanging bound GDP for GTP. It may form a complex with G proteins and stimulate Rho-dependent signals. This protein is activated by PI3-kinase. Mutations in this gene can cause X-chromosomal non-specific mental retardation. [provided by RefSeq]
Sequence: MNPEEQIVTWLISLGVLESPKKTICDPEEFLKSSLKNGVVLCKLINRLMPGSVEKFCLDPQTEADCINNINDFLKGCATLQVEIFDPDDLYSGVNFSKVLSTLLAVNKATEDQLSERPCGRSSSLSAANTSQTNPQGAVSSTVSGLQRQSKTVEMTENGSHQLIVKARFNFKQTNEDELSVCKGDIIYVTRVEEGGWWEGTLNGRTGWFPSNYVREIKSSERPLSPKAVKGFETAPLTKNYYTVVLQNILDTEKEYAKELQSLLVTYLRPLQSNNNLSTVEVTSLLGNFEEVCTFQQTLCQALEECSKFPENQHKVGGCLLSLMPHFKSMYLAYCANHPSAVNVLTQHSDELEQFMENQGASSPGILILTTNLSKPFMRLEKYVTLLQELERHMEDTHPDHQDILKAIVAFKTLMGQCQDLRKRKQLELQILSEPIQAWEGEDIKNLGNVIFMSQVMVQYGACEEKEERYLMLFSNVLIMLSASPRMSGFIYQGKIPIAGTVVTRLDEIEGNDCTFEITGNTVERIVVHCNNNQDFQEWLEQLNRLIRGPASCSSLSKTSSSSCSAHSSFSSTGQPRGPLEPPQIIKPWSLSCLRPAPPLRPSAALGYKERMSYILKESSKSPKTMKKFLHKRKTERKPSEEEYVIRKSTAALEEDAQILKVIEAYCTSANFQQGHGSSTRKDSIPQVLLPEEEKLIIEETRSNGQTIMEEKSLVDTVYALKDEVRELKQENKRMKQCLEEELKSRRDLEKLVRRLLKQTDECIRGESSSKTSILPSpecifications
NP_004831.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
113.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COOL2/Cool-2/KIAA0006/MRX46/PIXA/alpha-PIX/alphaPIX | |
ARHGEF6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9459 | |
ARHGEF6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ARHGEF6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.