Learn More
Cellular Engineering Technologies Cryopreserved vial of human amniotic membrane-derived mesenchymal stem cells (500,000 cells)
Supplier:  Cellular Engineering Technologies CR1006500
Encompass_Preferred
Cells harvested from amniotic membrane collected from a single adult human donor and then passaged three times to assure consistency. Amniotic fluid-derived mesenchymal stem cells (AF-MSCs) are adult, fibroblast-like, self-renewable, multipotent stem cells. These cells have significant differentiation capacity and immunomodulatory characteristics. These cells can generate mesodermal origin cells (chondrocytes, osteocytes, and adipocytes), neural cells, hepatocytes, epithelial cells, cardiomyocytes, and germ cells.These cells can be used for various cell therapy, immunomodulation, and regenerative medicine applications.Vial contains approximately 500,000 cells. Shipped with dry ice.
Catalog No. NC2753011
Explore available promotions