Learn More
Bon Opus Biosciences Recombinant Human AuroraA (122-403, N-His) - Source: E. coli
Supplier: Bon Opus Biosciences BP27750UG
Recombinant Human AuroraA (122-403) is produced by our mammalian expression system and the target gene encoding Glu122-Ser403 is expressed with a His tag at the N terminus. Accession#O14965. Formulation: Supplied as a 0.2 um filtered solution of 50mM Tris, 100mM NaCl, 10%glycerol, 1mM DTT, pH7.4