Learn More
Perkin Elmer US LLC HTS Sample Delivery Tubing and Loop Kit
Supplier: Perkin Elmer US LLC N8150145
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
PerkinElmers High Throughput System (HTS) affords ultra-fast analysis, dramatically reducing laboratory overheads and the cost of analysis. Using a metal-free fluid path, the system is able to quickly deliver the sample to plasma, while having uncompromised precision and detection limits. This kit includes: 0.75 mm I.D. Carrier Tubing, Qty. 1 (N8152922) 0.75 mm I.D. Internal Standard Tubing, Qty. 1 (N8152923) 0.75 mm I.D. Internal Standard (Port 7) Tubing, Qty. 1 (N8152924) 0.75 mm I.D. Carrier (Port 4) Tubing, Qty. 1 (N8152925) 1.5 mm I.D. Vacuum Pump (Port 2) Tubing, Qty. 1 (N8152926) 1.5 mm I.D. Waste Tubing, Qty. 1 (N8152927) 1 mL Sample Loop, 1.0 mm I.D., Qty. 1 (N8152929) CTFE Low Pressure Plug, 1/4-28 Fitting, Qty. 1 (09220596)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.