Learn More
Wilmad Labglass HT MNTL HEMI 300ML
Supplier: Wilmad Labglass LG8800110
Heating Mantle, Hemispherical. Size: 300mL. Watts: 180. Heating mantle features a soft, all glass fabric construction which supports the bottom half of the flask to allow full view of the contents of single or multi-neck flasks.
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov