Learn More
Wilmad Labglass HT MNTL HEMI 3000ML
Supplier: Wilmad Labglass LG8800118
Heating Mantle, Hemispherical. Size: 3000mL. Watts: 500. Heating mantle features a soft, all glass fabric construction which supports the bottom half of the flask to allow full view of the contents of single or multi-neck flasks.