Learn More
Wilmad Labglass HT MNTL BENCH TOP HEMI 22L
Supplier: Wilmad Labglass LG8875124
Heating Mantle, Hemispherical, Aluminum Housing. Size: 22000mL; Maximum Temperature: 450degreeC; Watts: 2 x 770; Volts: 115. Bench top, aluminum-housed heating mantle for round bottom flasks. Supplied w/ 4 foot long 3-Wire type SJT cord.