Learn More
Wilmad Labglass HT MNTL BENCH TOP HEMI 1L
Supplier: Wilmad Labglass LG8875114
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Heating Mantle, Hemispherical, Aluminum Housing. Size: 1000mL; Maximum Temperature: 450degreeC; Watts: 380; Volts: 115. Bench top, aluminum-housed heating mantle for round bottom flasks. Supplied w/ 4 foot long 3-Wire type SJT cord.
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov