Learn More
Leap PAL Parts Headspace Magnetic Crimp Cap 20mm, 5mm hole, PTFE/Clear Silicone, 3mm, 100 Pack
Supplier: Leap PAL Parts XAR20038368
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Cap, Crimp, Headspace, Magnetic With 5mm hole, 0.005 in. PTFE/Clear Silicone - 3.2mm, 100 Pack