Learn More
Advanced ImmunoChemical, Inc. Human Papillomavirus L1 Protein (HPVL1), Recombinant, Type 18, Analysis:Purity > 90 %
Supplier: Advanced ImmunoChemical, Inc. 8HPVL180.1MG
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Antigens/ Infectious Diseases / Human Papillomavirus L1 protein (HPVL1), recombinant, type 18. Analysis:Purity > 90 % (SDS-PAGE).Each lot is tested for protein content (Lowry method), lipid content (Total lipid), purity (SDS-PAGE), carbohydrate content (Anthrone method), antigen content (Immunoblot).Immunological activity:Tested in quantitative immunoblot with anti-HPVL1 antibodies.Purification:Purification is conducted from broken yeast cells by following scheme: clarification, ammonium sulfate precipitation, ultrafiltration, cation exchange chromatography, zonal centrifugation, size-exclusion chromatography, sterile filtration.Presentation:Presentation: 20 mM KH2PO4 (pH 7.2), 500 mM NaCl, 1.7 mM EDTA, 0.01% Tween-80, 1 mM 2-MercaptoethanoStorage Conditions:+ 4 °C (between +2 and +8 °C allowed)Material safety note:This product is sold for research use only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.