Learn More
Waters Corp HPLC HT with UV Standards Kit
Supplier: Waters Corp 700003742
Alliance HPLC with UV Detector standards kit for system assessment.12 Ampules (10ml standard solutions contain):- Solution HT 1 Qty 1 0.030 mg/mL Uracil ± 1% with 0.0100 mg/mL Caffeine ± 0.5%- Solution HT 2 Qty 1 0.030 mg/mL Uracil ± 1% with 0.0200 mg/mL Caffeine ± 0.5%- Solution HT 3 Qty 1 0.030 mg/mL Uracil ± 1% with 0.0300 mg/mL Caffeine ± 0.5%- Solution HT 4 Qty 1 0.030 mg/mL Uracil ± 1% with 0.0500 mg/mL Caffeine ± 0.5%- Solution HT 5 Qty 3 0.030 mg/mL Uracil ± 1% with 0.0600 mg/mL Caffeine ± 0.5%- Solution HT 6 Qty 2 0.010 mg/mL Uracil ± 1%- Solution HT 7 Qty 1 0.40 ug/mL Caffeine ± 0.5%- Solution HT 8 Qty 1 4.00 mg/mL Caffeine ± 0.5%- Solution HT 9 Qty 1 Blank (mobile phase only)Caffeine and Uracil > 99%All standards are in 30% HPLC Grade Methanol and 70% 18M ASTM Type 1 Lab Water, V/VCertificate of Analysis
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.