Learn More
Air Science Usa Horizontal Clean Bench 24" Wide with UV System
Supplier: Air Science Usa HLF24A(UV)
Horizontal Laminar Flow Cabinet 24" Wide With UV Lamp and front cover. Protects your non-hazardous applications with horizontal unidirectional (laminar) ISO Class 4 air across the work area. Includes Stainless Steel Workurface. CART-BW-HLF-24 Base Stand avilable and sold separatly.