Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hourglass International™ HandPRO™ AirSoft900™ Nitrile Exam Gloves

Click to view available options
Size:
Large
Medium
Small
X-Large
X-Small
Description
- Powder free
- Not made with natural rubber latex
- Textured fingertips
- Static dissipative ESD qualities
- Tested safe for use with chemotherapy drugs
Specifications
Certifications/Compliance | ASTM D6319, ASTM D6978-05, ASTM F1671, FDA 21 CFR 177.2600, USP 800 |
Certifications/Compliance (Europe) | EN 455 |
Chemotherapeutic Rated | Yes |
Accelerator Free | No |
Applications | Healthcare, Laboratory, Industrial |
AQL | 1.5 |
Color | Blue |
Disposable | Yes |
Exterior Finish | Partially Textured |
Finger Thickness (English) | 5.5 mil |
Show More |