Learn More
Bellco Glass, Inc. Hot Shaker Plus 115V
Supplier: Bellco Glass, Inc. 774622110
Bellco’s Sci-ERA Hot Shaker Plus is a slow speed, large orbit shaker for the incubation of electrophoretic gels and filter hybridization media. Stringent temperature control with a sensitivity of +/-0.1oC is useful for many biochemical assays. This Bellco