Learn More
Innovative Research Inc Horse Red Blood Cells Packed 100% 100ml
Supplier:  Innovative Research Inc IHSRBC100P100ML
Encompass_Preferred
Horse Red Blood Cells from Innovative Research are manufactured using whole blood collections from horses. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 100% suspension in phosphate buffered saline (PBS). Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4877