Learn More
Rockland Immunochemicals Horse Blood In Sodium Citrate 50 mL
SDP
Supplier:  Rockland Immunochemicals R2070050
Encompass_Preferred
Citrated horse blood is aseptically collected and prepared to order from our colony of animals. Citrated horse blood is ideal for blood agar plate preparations. Blood Agar is a general purpose enriched medium often used to grow fastidious organisms and to differentiate bacteria based on their hemolytic properties. Citrated Horse RBCs are also useful for the titration of complement adsorption procedures testing for agglutinins/HA assays and for the preparation of stroma as particulate reagents. Horse red blood cells are perishable and are collected and processed upon receipt of your order.
Catalog No. 50-194-3982