Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CBS Scientific Horizontal Standard Midi-Gel Kits
Used with Midi Horizontal System.
Supplier: CBS Scientific MHU602PK
Description
CBS Midi Horizontal Productivity Packages feature platinum electrode cassettes which provide rapid sample velocity and clear focused bands without smiling.- Platinum electrode cassettes can be easily removed for cleaning or replacement
- High quality, heavy duty construction with vast array of accessory options
- Winged gel trays for easy handling. No more sticking fingers into slippery buffer to extract gel tray
- Addition of external gel caster and additional gel tray allows researcher to use time more efficiently
- MHU-602 Midi-Horizontal runs up to a full microtitre plate (108 samples) per gel
- Tapered baffles allow easy in situ gel formation with or without gel tray
- Run up to 32 samples per gel. Two slots in each gel tray
- Combs are printed with catalog number and thickness for easy identification
- Choice of double-sided adjustable height combs gives researcher more options
- Optional PTFE-coated aluminum combs for low-friction comb removal and accelerated well formation
- Horizontal Midi-Gels are available in four vivid colors; green, yellow, blue or fluorescent pink
- Color-coded horizontal systems are recommended to differentiate exclusive use by specific lab or for specific purpose
Specifications
CE | |
Compatible Gel Tray: WGT-600 | |
L x W: 16 x 14 cm (6.2 x 5.5 in.) | |
3 Years | |
27.5 x 16.7 x 14 cm (10.8 x 6.5 x 5.5 in.) |
Pink | |
Midi-Gel Kit | |
750 mL | |
Gel Tray I.D.: 16 x 12.7 cm (6.2 x 5 in.) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction