Learn More
Expotech Usa Inc Horiba S-316 Solvent, 1.5 kg
Supplier: Expotech Usa Inc 1459145
Horiba OCMA Analyzers use this S-316 extraction solvent to extract the oil components from oily water samples, soil samples, or product surfaces. The extract is analyzed using IR absorbance, a non-dispersive infrared spectrophotometric technique which is specific to hydrocarbons such as oil.