Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HORIBA ABX Pentra™ 400 Clinical Chemistry Analyzer Controls
$386.00 - $651.50
Specifications
Certifications/Compliance | CE |
---|---|
Format | Lyophilized |
For Use With (Equipment) | ABX Pentra™ C400 |
Intended Use | Serum, Plasma, Whole Blood or Hemolysate (HbA1c), and Urine (Microalbumin) |
Catalog Number | Mfr. No. | Type | Format | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Format | Quantity | Price | Quantity & Availability | |||
23-600-412
|
HORIBA
1220001621 |
Immuno I Control L/H | Lyophilized | 1 x 3 mL + 1 x 3 mL |
Each for $651.50
|
|
23-600-413
|
HORIBA
1220001622 |
Immuno II Control L/H | Lyophilized | 1 x 3 mL + 1 x 3 mL |
Each for $568.00
|
|
23-600-501
|
HORIBA
1220001786 |
CK Control | Lyophilized | 4 x 3 mL |
Each for $386.00
|
|
23-600-443
|
HORIBA
1220001654 |
P Control | Lyophilized | 10 x 5 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Specifications
CE | |
Lyophilized | |
ABX Pentra™ C400 | |
Serum, Plasma, Whole Blood or Hemolysate (HbA1c), and Urine (Microalbumin) |
Safety and Handling
ShelfLife : 5 Years
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title