Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HORIBA ABX Lysebio
For Pentra 60 C+ and Pentra XL80
$254.00 - $364.00
Specifications
For Use With (Equipment) | Pentra 60 C+ and Pentra XL80 |
---|---|
Type | Lysebio Reagent |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-450-100
|
HORIBA
1210906013 |
400 mL |
Each for $254.00
|
|
|||||
23-450-102
|
HORIBA
1210906012 |
1 L |
Each for $364.00
|
|
|||||
Specifications
Pentra 60 C+ and Pentra XL80 | |
Lysebio Reagent |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title