Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ PRO 5050 Struximity Leather Boots, Berry-Compliant, Wide
Lightweight, comfortable, high-speed, wide-width leather pull-on bunker boots dual certified for both Structural and Proximity (ARFF) firefighting
Specifications
Liner Material | Cambrelle |
---|---|
Material | Leather, Dupont Kevlar Aramid Fiber, CrossTech |
Weight (English) | 6.6 lb. |
Description
Made of high performance fire resistant and waterproof leather and Goodyear welt made to withstand the harsh proximity environment
- Athletic construction delivers unencumbered performance and agility
- Full height bootie combines an aluminized radiant shield, 7.5 oz Kevlar thermal felt quilted to Cambrelle™ liner and laminated to Crosstech™ Moisture barrier fabric for protection from heat, fluids, and internal abrasion
- Specially designed profiled boot shaft is large enough to accommodate large calves while still fitting comfortably under proximity pants
- Vibram™ lug replaceable soles with Fire and Ice compound are cemented and security stitched using Goodyear welt construction for superior durability protection
- Fire and Ice sole compound improves winter traction on ice and snow
- Steel bottom plate and shank offer protection from puncture and dangerous twisting
- Improved heel pocket and comfortable molded insert defend against heel slippage
- APT Barcode — Departmental NFPA 1851 Tracking
- Certified to NFPA 1971, current edition for Proximity and Structural Fire Fighting
- Weight: 6.6 pounds
- Wide width
- Union made in USA. All components are Berry Amendment compliant
Specifications
Cambrelle | |
Leather, Dupont Kevlar Aramid Fiber, CrossTech | |
6.6 lb. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title