Learn More
Honeywell™ PRO 5050 Struximity Leather Boots, Berry Compliant
Lightweight, comfortable, high-speed, leather pull-on bunker boots dual certified for both Structural and Proximity (ARFF) firefighting
Specifications
Liner Material | Cambrelle |
---|---|
Material | Leather, CrossTech, Dupont Kevlar Aramid Fiber |
Description
Made of high performance fire resistant and waterproof leather and Goodyear welt made to withstand the harsh proximity environment
- Athletic construction delivers unencumbered performance and agility
- Full height bootie combines an aluminized radiant shield, 7.5 oz Kevlar thermal felt quilted to Cambrelle™ liner and laminated to Crosstech™ Moisture barrier fabric for protection from heat, fluids, and internal abrasion
- Specially designed profiled boot shaft is large enough to accommodate large calves while still fitting comfortably under proximity pants
- Vibram™ lug replaceable soles with Fire and Ice compound are cemented and security stitched using Goodyear welt construction for superior durability protection
- Fire and Ice sole compound improves winter traction on ice and snow-even on wet ice
- Steel bottom plate and shank offer protection from puncture and dangerous twisting
- Improved heel pocket and comfortable molded insert defend against heel slippage
- APT Barcode — Departmental NFPA 1851 Tracking
- Certified to NFPA 1971, current edition for Proximity and Structural Fire Fighting
- Weight: 6.6 pounds
- Union made in USA
- All components are Berry Amendment compliant
Specifications
Cambrelle | |
Leather, CrossTech, Dupont Kevlar Aramid Fiber |