Learn More
Honeywell™ Honeywell™ North™ Silver Shield Chemical Resistant Sleeves
Click to view available options
Sleeves; 20 in. L; One Size Fits All
Description
- Five layer PE and EVOH constructions provides excellent chemical resistant to over 280 different chemicals including alchohols, aliphatics, aromatice, chlorines, ketones and esters
Specifications
Product Type | Sleeves |
Material | Composite Fabric |
Cuff Style | Elastic |