Learn More
Honeywell Analytics™ GasAlertQuattro Rechargable Multigas Detectors
$1708.00 - $2170.00
Specifications
Alarm Types | Audible, visual, vibrating |
---|---|
Battery Life | 18 to 20 hr. |
Depth (English) | 1.87 in. |
Depth (Metric) | 4.7 cm |
Height (English) | 5.12 in. |
Catalog Number | Mfr. No. | Color | Resolution | Components Measured | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Resolution | Components Measured | Price | Quantity & Availability | |||
19-413-304
|
Honeywell Analytics™
QTXWHMRBNA |
Black | O2: 0.1%, LEL: 1%, H2S: 1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide, LEL, Oxygen |
Each for $2,170.00
|
|
|||
19-413-303
|
Honeywell Analytics™
QTXWHMRYNA |
Yellow | H2S: 0.1ppm, CO: 1ppm, O2: 0.1%, Combustible gases: 1%, 0.1% | Carbon Monoxide, Hydrogen Sulfide, LEL, Oxygen |
Each for $2,170.00
|
|
|||
19-413-319
|
Honeywell Analytics™
QTXWH0RYNA |
Yellow | H2S: 0.1ppm, O2: 0.1%, Combustible gases: 1%, 0.1% | Carbon Monoxide, Hydrogen Sulfide, LEL |
Each for $2,050.00
|
|
|||
19-413-320
|
Honeywell Analytics™
QTXW0MRYNA |
Yellow | CO: 1ppm, O2: 0.1%, Combustible gases: 1%, 0.1% | Carbon Monoxide, LEL, Oxygen |
Each for $2,050.00
|
|
|||
19-413-321
|
Honeywell Analytics™
QTX0HMRYNA |
Yellow | O2: 0.1%, H2S: 1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide, Oxygen |
Each for $2,050.00
|
|
|||
19-413-324
|
Honeywell Analytics™
QTX0H0RYNA |
Yellow | O2: 0.1%, H2S: 1ppm | Hydrogen Sulfide, Oxygen |
Each for $1,926.00
|
|
|||
19-413-323
|
Honeywell Analytics™
QTXW00RYNA |
Yellow | O2: 0.1%, LEL: 1% | LEL, Oxygen |
Each for $1,926.00
|
|
|||
19-413-326
|
Honeywell Analytics™
QTX00MRYNA |
Yellow | O2: 0.1%, CO: 1ppm | Carbon Monoxide, Oxygen |
Each for $1,926.00
|
|
|||
19-413-328
|
Honeywell Analytics™
QT00HMRYNA |
Yellow | H2S: 0.1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide |
Each for $1,926.00
|
|
|||
19-413-347
|
Honeywell Analytics™
QTXWH0RBNA |
Black | O2: 0.1%, LEL: 1%, H2S: 1ppm | Hydrogen Sulfide, LEL, Oxygen |
Each for $2,050.00
|
|
|||
19-413-348
|
Honeywell Analytics™
QTXW0MRBNA |
Black | O2: 0.1%, LEL: 1%, CO: 1ppm | Carbon Monoxide, LEL, Oxygen |
Each for $2,050.00
|
|
|||
19-413-349
|
Honeywell Analytics™
QTX0HMRBNA |
Black | O2: 0.1%, H2S: 1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide, Oxygen |
Each for $1,708.00
|
|
|||
19-413-406
|
Honeywell Analytics™
QT00HMRBNA |
Black | H2S: 1ppm, CO: 1ppm | Carbon Monoxide, Hydrogen Sulfide |
Each for $1,926.00
|
|
|||
19-413-404
|
Honeywell Analytics™
QTX00MRBNA |
Black | O2: 0.1%, CO: 1ppm | Carbon Monoxide, Oxygen |
Each for $1,926.00
|
|
|||
19-413-401
|
Honeywell Analytics™
QTXW00RBNA |
Black | O2: 0.1%, LEL: 1% | LEL, Oxygen |
Each for $1,926.00
|
|
|||
19-413-402
|
Honeywell Analytics™
QTX0H0RBNA |
Black | O2: 0.1%, H2S: 1ppm | Hydrogen Sulfide, Oxygen |
Each for $1,926.00
|
|
|||
Description
Specifications
Audible, visual, vibrating | |
1.87 in. | |
5.12 in. | |
10 to 100% | |
-20°C to +50°C | |
316g | |
8.25 cm |
18 to 20 hr. | |
4.7 cm | |
13 cm | |
-4°F to +122°F | |
11.15 oz. | |
3.25 in. |