Learn More
Honeywell Analytics™ GasAlertQuattro Confined Space Kit
Supplier: Honeywell Analytics™ QTXWHMRYNACS
Description
Specifications
Audible, visual, vibrating | |
1.87 in. | |
Carbon Monoxide, Hydrogen Sulfide, LEL, Oxygen | |
13 cm | |
O2: 0.1%, CO: 1ppm, H2S: 0.1ppm, LEL: 1% | |
-20°C to +50°C | |
338g | |
8.2 cm |
2 Years | |
4.7 cm | |
5.12 in. | |
10 to 100% | |
-4°F to +122°F | |
11.92 oz. | |
3.25 in. |