Learn More
Honeywell Analytics™ GasAlert Extreme Single-Gas Detectors
For any single gas hazard, choose from a wide range of available toxic gas models. The GasAlert Extreme Single-Gas Detector provides a simple, low-cost option requiring minimal training. Its compact, lightweight design makes it versatile and comfortable.
$982.50 - $1564.00
Specifications
Certifications/Compliance | ATEX: CE Ex II 1 G Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, Class II, Division 1, Group E, F, G, IECEx: Exia IIC T4, INMETRO: BR-Exia IIC T4, EMC Directive 89/336/EEC |
---|---|
Depth (English) | 1.1 in. |
Depth (Metric) | 2.8 cm |
Height (English) | 3.75 in. |
Height (Metric) | 9.52 cm |
Catalog Number | Mfr. No. | Components Measured | Humidity Range | Measurement Range | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Components Measured | Humidity Range | Measurement Range | Price | Quantity & Availability | |||
19-413-358
|
Honeywell Analytics™
GAXTEDL |
Ethylene Oxide | 15 to 90% | 0 to 100ppm |
Each for $1,564.00
|
|
|||
19-413-360
|
Honeywell Analytics™
GAXTHDL |
Hydrogen Sulfide | 15 to 90% | 0 to 100ppm |
Each for $1,060.00
|
|
|||
19-413-363
|
Honeywell Analytics™
GAXTPDL |
Phosphine | 15 to 90% | 0 to 5ppm |
Each for $1,080.00
|
|
|||
19-413-365
|
Honeywell Analytics™
GAXTVDL |
Chlorine Dioxide | 15 to 95% | 0 to 1ppm |
Each for $982.50
|
|
|||
19-413-359
|
Honeywell Analytics™
GAXTGDL |
Ozone | 15 to 90% | 0 to 1ppm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
19-413-356
|
Honeywell Analytics™
GAXTCDL |
Chlorine | 10 to 95% | 0 to 50ppm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
19-413-357
|
Honeywell Analytics™
GAXTDDL |
Nitrogen Dioxide | 15 to 95% | 0 to 99.9ppm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
19-413-362
|
Honeywell Analytics™
GAXTNDL |
Nitric Oxide | 15 to 90% | 0 to 250ppm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
19-413-368
|
Honeywell Analytics™
GAXTA2DL |
Ammonia | 15 to 90% | 0 to 400ppm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
19-413-364
|
Honeywell Analytics™
GAXTSDL |
Sulfur Dioxide | 15 to 90% | 0 to 100ppm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
19-413-367
|
Honeywell Analytics™
GAXTH2DL |
Hydrogen Sulfide | 15 to 90% | 0 to 500ppm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
19-413-366
|
Honeywell Analytics™
GAXTZDL |
Hydrogen Cyanide | 15 to 95% | 0 to 30ppm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
For any single gas hazard, choose from a wide range of available toxic gas models. TheGasAlert Extreme Single-Gas Detector provides a simple, low-cost option requiringminimal training. Its compact, lightweight design makes it versatile and comfortable.
- Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms on start up
- Secure and accurate history with datalogging with passcode protection
- Continuous LCD shows real-time gas concentrations
- Simple automatic calibration procedure
- Equipped with internal vibrating alarm for high noise areas
- Visual, vibrating, audible alarms (95 dB) - Low, High, TWA and STEL
- Replaceable 3V battery
- Water-resistant
- Bright wide-angled visual alarm bars
- Two-year field-replaceable battery (1.5 year life) and sensor
- Adjustable alarm setpoints
- Multilanguage display options
- Compatible with Sampler motorized pump
- Tests: Sensor integrity, circuitry, battery and audible/visual alarms on activation, battery
- Humidity Range: 15% to 90% for all but O2 (0% to 99%) and Cl2 (10% to 95%) sensors
- Size: 2.8 × 5.0 × 9.5cm (1.1 × 2.0 × 3.75 in.)
- Weight: 82g (2.9 oz.)
Applications:
For single-gas detection of H2S, SO2, HCN, CO, NH3, Cl2, NO, NO2, PH3, ETO, ClO2, O3, O2. (See Product Type at right.)
Warranty and Services
Two years including sensor, except Ammonia (one year)
Specifications
ATEX: CE Ex II 1 G Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, Class II, Division 1, Group E, F, G, IECEx: Exia IIC T4, INMETRO: BR-Exia IIC T4, EMC Directive 89/336/EEC | |
2.8 cm | |
9.52 cm | |
2.9 oz. | |
2 in. |
1.1 in. | |
3.75 in. | |
2 Years | |
83g | |
5.08 cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.