Learn More
Honeywell Analytics™ Auxiliary Pump Filter
For use with GasAlertMicro 5 Series portable gas monitors.
Supplier: Honeywell Analytics™ M5AFK3
Description
- Use with Generation 2 pump modules (with filter window)
- 5 per Kit
Specifications
GasAlertMicro 5 Series Portable Gas Monitors | |
Auxiliary Pump Filter | |
Use with Generation 2 pump modules (with filter window) |