Learn More
Honeywell Analytics™ Bump Test Kit
Accessory for E3Point Toxic and Combustible Gas Monitor.
Supplier: Honeywell Analytics™ TEST1A
Description
For E3Point Toxic and Combustible Gas Monitor.Specifications
E3 Point Toxic and Combustible Gas Monitor | |
Bump Test Kit | |
Balloon adapter and quad gas cylinder: H2S (40 ppm), CO (200 ppm), CH4 (2.5%), O2 (10%) |